Thermo Scientific Nalgene Straight-Side, Wide-Mouth Jars, PP
Choose clear polycarbonate or for better chemical resistance, translucent polypropylene. All containers include polypropylene screw closures. Closures are grooved for easy stacking. Autoclavable (autoclave pieces separately).
8,500.00 - ₹23,100.00INR / Pkg of 12
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene 2118-9050 Straight-Sided Polypropylene Wide-Mouth Jar, 15 mL 0675848 Mfr # 2118-9050
COMPARE
15 38 mm Polypropylene(PP)
₹8,500.00INR / Pkg of 12
Thermo Scientific Nalgene 2118-0001 Straight-Sided Polypropylene Wide-Mouth Jar, 30 mL 0675850 Mfr # 2118-0001
COMPARE
30 43 mm Polypropylene(PP)
₹9,100.00INR / Pkg of 12
Thermo Scientific Nalgene 2118-0002 Straight-Sided Polypropylene Wide-Mouth Jar, 60 mL 0675852 Mfr # 2118-0002
COMPARE
60 53 mm Polypropylene(PP)
₹10,900.00INR / Pkg of 12
Thermo Scientific Nalgene 2118-0004 Straight-Sided Polypropylene Wide-Mouth Jar, 125 mL 0675854 Mfr # 2118-0004
COMPARE
125 70 mm Polypropylene(PP)
₹17,200.00INR / Pkg of 12
Thermo Scientific Nalgene 2118-0008 Straight-Sided Polypropylene Wide-Mouth Jar, 250 mL 0675856 Mfr # 2118-0008
COMPARE
250 70 mm Polypropylene(PP)
₹10,300.00INR / Pkg of 6
Thermo Scientific Nalgene 2118-0016 Straight-Sided Polypropylene Wide-Mouth Jar, 500 mL 0675858 Mfr # 2118-0016
COMPARE
500 120 mm Polypropylene(PP)
₹16,300.00INR / Pkg of 6
Thermo Scientific Nalgene 2118-0032 Straight-Sided Polypropylene Wide-Mouth Jar, 1000 mL 0675860 Mfr # 2118-0032
COMPARE
1000 120 mm Polypropylene(PP)
₹23,100.00INR / Pkg of 6
Showing 1- 7 of 7