Thermo Scientific Nalgene Graduated Cylinders, PMP
Easy-to-read raised graduations
  • Chemical and heat resistant
  • Large pour spout
  • Autoclavable
Transparent polymethylpentene (PMP) cylinders with large pour spout offer good chemical and heat resistance. Easy-to-read, raised graduations allow precise measuring.
2,100.00 - ₹9,100.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Thermo Scientific Nalgene 3665-0010 graduated polymethylpentene cylinder, 10 mL WW-06134-15 Mfr # 3665-0010
COMPARE
10 0.2 Polymethylpentene
₹2,100.00INR / Each
Thermo Scientific Nalgene 3665-0025 graduated polymethylpentene cylinder, 25 mL WW-06134-25 Mfr # 3665-0025
COMPARE
25 0.5 Polymethylpentene
₹2,600.00INR / Each
Thermo Scientific Nalgene 3665-0050 graduated polymethylpentene cylinder, 50 mL WW-06134-35 Mfr # 3665-0050
COMPARE
50 1 Polymethylpentene
₹3,300.00INR / Each
Thermo Scientific Nalgene 3665-0100 graduated polymethylpentene cylinder, 100 mL WW-06134-45 Mfr # 3665-0100
COMPARE
100 1 Polymethylpentene
₹3,900.00INR / Each
Thermo Scientific Nalgene 3665-0250 graduated polymethylpentene cylinder, 250 mL WW-06134-55 Mfr # 3665-0250
COMPARE
250 2 Polymethylpentene
₹5,200.00INR / Each
Thermo Scientific Nalgene 3665-0500 graduated polymethylpentene cylinder, 500 mL WW-06134-65 Mfr # 3665-0500
COMPARE
500 5 Polymethylpentene
₹7,300.00INR / Pkg of 1
Thermo Scientific Nalgene 3665-1000 graduated polymethylpentene cylinder, 1000 mL WW-06134-75 Mfr # 3665-1000
COMPARE
1000 10 Polymethylpentene
₹9,100.00INR / Each
Showing 1- 7 of 7