Thermo Scientific Finnpipette F3 Pipettors
Enhanced accuracy and precision
  • Advanced volume gearing
  • Large and clear display
  • Light pipetting action
  • Double-action pipetting hat
  • Autoclavable tip cone and tip ejector
Finnpipette F3 pipettors meet your high standards of accuracy and precision. An innovative, modular volume adjustment mechanism not only allows 50 clicks per revolution but is separated from the pipettor hull for increased durability. A sizeable black and white display with large digits provides you with easy volume recognition. Additionally the Finnpipette F3 has been designed to increase comfort and ergonomics. The double-action pipetting hat incorporates a rotating upper part to enable smooth and steady pipetting action without accidental volume alterations.
13,100.00 - ₹14,400.00INR / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Max Volume (mL) Min Volume (mL)
Availability Pricing
Thermo Scientific Finnpipette 4640000 Variable Volume Pipettor, 1 to 10 µL micro-tip 2599900 Mfr # 4640000
COMPARE
Yes 0.01 0.001
₹14,400.00INR / Each
Thermo Scientific Finnpipette 4640010 Variable Volume Pipettor, 1 to 10 µL universal-tip 2599901 Mfr # 4640010
COMPARE
Yes 0.01 0.001
₹14,400.00INR / Each
Thermo Scientific Finnpipette 4640020 Variable Volume Pipettor, 2 to 20 µL micro-tip 2599902 Mfr # 4640020
COMPARE
Yes 0.02 0.002
₹14,400.00INR / Each
Thermo Scientific Finnpipette 4640030 Variable Volume Pipettor, 2 to 20 µL universal-tip 2599903 Mfr # 4640030
COMPARE
Yes 0.02 0.002
₹14,400.00INR / Each
Thermo Scientific Finnpipette 4640040 Variable Volume Pipettor, 10 to 100 µL 2599904 Mfr # 4640040
COMPARE
Yes 0.1 0.01
₹13,100.00INR / Each
Thermo Scientific Finnpipette 4640050 Variable Volume Pipettor, 20 to 200 µL 2599905 Mfr # 4640050
COMPARE
Yes 0.2 0.02
₹14,400.00INR / Each
Thermo Scientific Finnpipette 4640060 Variable Volume Pipettor, 100 to 1000 µL 2599906 Mfr # 4640060
COMPARE
Yes 1 0.1
₹13,100.00INR / Each
Thermo Scientific Finnpipette 4640070 Variable Volume Pipettor, 1 to 10 mL 2599907 Mfr # 4640070
COMPARE
Yes 10 1
₹13,100.00INR / Each
Thermo Scientific Finnpipette 4650000 Fixed Volume Pipettor, 1 µL micro-tip 2599910 Mfr # 4650000
COMPARE
Yes 0.001
Call For Price
Thermo Scientific Finnpipette 4650010 Fixed Volume Pipettor, 5 µL micro-tip 2599911 Mfr # 4650010
COMPARE
Yes 0.005
Call For Price
Thermo Scientific Finnpipette 4650020 Fixed Volume Pipettor, 10 µL 2599912 Mfr # 4650020
COMPARE
Yes 0.01
Call For Price
Thermo Scientific Finnpipette 4650030 Fixed Volume Pipettor, 25 µL 2599913 Mfr # 4650030
COMPARE
Yes 0.025
Call For Price
Thermo Scientific Finnpipette 4650040 Fixed Volume Pipettor, 50 µL 2599914 Mfr # 4650040
COMPARE
Yes 0.05
Call For Price
Thermo Scientific Finnpipette 4650050 Fixed Volume Pipettor, 100 µL 2599915 Mfr # 4650050
COMPARE
Yes 0.1
Call For Price
Thermo Scientific Finnpipette 4650060 Fixed Volume Pipettor, 250 µL 2599916 Mfr # 4650060
COMPARE
Yes 0.25
Call For Price
Thermo Scientific Finnpipette 4650070 Fixed Volume Pipettor, 500 µL 2599917 Mfr # 4650070
COMPARE
Yes 0.5
Call For Price
Thermo Scientific Finnpipette 4650080 Fixed Volume Pipettor, 1000 µL 2599918 Mfr # 4650080
COMPARE
Yes 1
Call For Price
Showing 1- 17 of 17