Thermo Scientific Barnstead NANOpure Water Purification Systems Cartridges
Keep systems operating precisely
  • Use with Nanopure®, Nanopure Infinity®, Easypure® II, E-Pure™, Bantam™, and B-Pure™ systems
  • Choose cartridge based upon your application
Cartridge kits include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagentgrade water. Kits that include organic-free cartridges produce water that has less than 10 ppb
organic carbon. Kits for Nanopure Infinity and Nanopure Dlamond systems include final filter; order final filter for quad-style Nanopure and E-Pure systems separately.
332,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description Compatible with
Availability Pricing
Thermo Scientific D50280 Organic-Free RO or Distilled Feed Cartridge Pack 9927750 Mfr # D50280
COMPARE
Organic-Free RO or Distilled Feed Cartridge Pack Thermo Scientific Barnstead NANOpure™
₹332,000.00INR / Each
Thermo Scientific Barnstead D50281 NANOpure Cartridge Pack, organic-free DI feed 9927752 Mfr # D50281
COMPARE
NANOpure Cartridge Pack, organic-free DI feed Thermo Scientific Barnstead NANOpure™
₹332,000.00INR / Each
Thermo Scientific Barnstead D50282 NANOpure Cartridge Packs, Type I RO and distilled feed 9927754 Mfr # D50282
COMPARE
NANOpure Cartridge Packs, Type I RO and distilled feed Thermo Scientific Barnstead NANOpure™
₹332,000.00INR / Each
Thermo Scientific D50283 Cartridge Pack for NANOpure Water Purification System 9927756 Mfr # D50283
COMPARE
Cartridge Pack for NANOpure Water Purification System Thermo Scientific Barnstead NANOpure™
₹332,000.00INR / Each