Thermo Scientific D50283 Cartridge Pack for NANOpure Water Purification System
Representative image only.
Thermo Scientific D50283 Cartridge Pack for NANOpure Water Purification System
Thermo Scientific | Mfr: D50283| Item: 9927756
Keep systems operating precisely
₹332,000.00INR / Each
₹332,000.00INR / Each
Product Description
Key Features
  • Use with Nanopure®, Nanopure Infinity®, Easypure® II, E-Pure™, Bantam™, and B-Pure™ systems
  • Choose cartridge based upon your application
More About this Item
Cartridge kits include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagentgrade water. Kits that include organic-free cartridges produce water that has less than 10 ppb
organic carbon. Kits for Nanopure Infinity and Nanopure Dlamond systems include final filter; order final filter for quad-style Nanopure and E-Pure systems separately.
Kit includes one organic/colloid absorption, three mixed bed, and one organic free.
Specifications
  • Compatible withThermo Scientific Barnstead NANOpure™
  • ApplicationsLow organic (non-detectable level)
  • DescriptionCartridge Pack for NANOpure Water Purification System