Thermo Scientific Barnstead D50282 NANOpure Cartridge Packs, Type I RO and distilled feed
Representative image only.
Thermo Scientific Barnstead D50282 NANOpure Cartridge Packs, Type I RO and distilled feed
Thermo Scientific | Mfr: D50282| Item: 9927754
Keep systems operating precisely
₹332,000.00INR / Each
₹332,000.00INR / Each
Product Description
Key Features
  • Use with Nanopure®, Nanopure Infinity®, Easypure® II, E-Pure™, Bantam™, and B-Pure™ systems
  • Choose cartridge based upon your application
More About this Item
Cartridge kits include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagentgrade water. Kits that include organic-free cartridges produce water that has less than 10 ppb
organic carbon. Kits for Nanopure Infinity and Nanopure Dlamond systems include final filter; order final filter for quad-style Nanopure and E-Pure systems separately.
Kit contains one organic absorption, three mixed bed, and one organic free.
Specifications
  • Compatible withThermo Scientific Barnstead NANOpure™
  • ApplicationsLow organic (non-detectable level)
  • DescriptionNANOpure Cartridge Packs, Type I RO and distilled feed