Thermo Scientific Barnstead Final Filters for Deionization Systems
Use with Nanopure®, Nanopure RO, Easypure® II, E-Pure™, and B-Pure™ systems
Final filters remove particulates down to 0.2 µm to provide bacteria- free water. Constructed of hollow fiber material, filters do not contain any wetting agents or surfactants. Filters are radiation sterilized and individually sealed.
Autoclavable.
51,800.00 - ₹97,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description Compatible with Max Flow Rate (LPM)
Availability Pricing
Thermo Scientific Barnstead D3750 NANOpure/EASYpure II Final Filter 9925600 Mfr # D3750
COMPARE
NANOpure/EASYpure II Final Filter E-Pure, Nanopure II, Nanopure Diamond, and Easypure II 2
₹51,800.00INR / Each
Thermo Scientific DISPENSER FINAL FILTER 9929927 Mfr # 09.1003
COMPARE
DISPENSER FINAL FILTER Nanopure®, Nanopure RO, Easypure® II, E-Pure™, and B-Pure™ systems
₹97,300.00INR / Each