Thermo Scientific Barnstead D3750 NANOpure/EASYpure II Final Filter
Representative image only.
Thermo Scientific Barnstead D3750 NANOpure/EASYpure II Final Filter
Thermo Scientific | Mfr: D3750| Item: 9925600
Use with Nanopure®, Nanopure RO, Easypure® II, E-Pure™, and B-Pure™ systems
₹51,800.00INR / Each
₹51,800.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Final filters remove particulates down to 0.2 µm to provide bacteria- free water. Constructed of hollow fiber material, filters do not contain any wetting agents or surfactants. Filters are radiation sterilized and individually sealed.
Autoclavable.
1 Year Warranty
Specifications
  • Compatible withE-Pure, Nanopure II, Nanopure Diamond, and Easypure II
  • Max Flow Rate (LPM)2
  • DescriptionNANOpure/EASYpure II Final Filter