Thermo Scientific DISPENSER FINAL FILTER
Representative image only.
Thermo Scientific DISPENSER FINAL FILTER
Thermo Scientific | Mfr: 09.1003| Item: 9929927
Use with Nanopure®, Nanopure RO, Easypure® II, E-Pure™, and B-Pure™ systems
₹97,300.00INR / Each
₹97,300.00INR / Each
Product Description
Key Features
  • 3 month warranty
More About this Item
Final filters remove particulates down to 0.2 µm to provide bacteria- free water. Constructed of hollow fiber material, filters do not contain any wetting agents or surfactants. Filters are radiation sterilized and individually sealed.
Autoclavable.
3 Month Warranty
Specifications
  • Compatible withNanopure®, Nanopure RO, Easypure® II, E-Pure™, and B-Pure™ systems
  • DescriptionDISPENSER FINAL FILTER