SI Analytics Lab 865 and Lab 875P pH Benchtop Meters
  • Rapid data transfer via USB interface
  • Manual or timer controlled data logging of measured values
  • Protocols include date, time, and ID (Lab 875P model) and sensors serial
  • numbers for GLP compliance
  • Excel Add-in included for the formatted output of all data and calibration
  • protocols
  • Output data directly to integrated printer (Lab 875P model)
232,000.00 - ₹344,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem Availability Pricing
COMPARE
₹232,000.00INR / Each
COMPARE
₹344,000.00INR / Each