Cole-Parmer LAB 875P BENCHTOP PH METER
Representative image only.
Cole-Parmer LAB 875P BENCHTOP PH METER
Cole-Parmer | Item: 1596163
₹344,000.00INR / Each
₹344,000.00INR / Each
Product Description
Includes
pH/temperature combination electrode (BlueLine 14 pH), calibration solutions, stand, USB cable, power supply, and four AA 1.5 V batteries.
Key Features
  • Rapid data transfer via USB interface
  • Manual or timer controlled data logging of measured values
  • Protocols include date, time, and ID (Lab 875P model) and sensors serial
  • numbers for GLP compliance
  • Excel Add-in included for the formatted output of all data and calibration
  • protocols
  • Output data directly to integrated printer (Lab 875P model)
More About this Item
Innovative Lab 875P Precision Multiparameter Meter with a digital measuring
channel is optimal in the world of digital multiparameter measurement using
IDS technology (refer page 310 for details). IDR technology allows optimized
measurements and efficient documentation in the simplest manner—
the meter supports automatic sensor recognition and the IDS sensors
can be moved from meter to meter without having to recalibrate.
This one-channel multiparameter meter lets you measure pH,
ORP, conductivity, and DO quickly and reliably. It offers automatic
digital sensor recognition, sensor quality control (QSC), and
continuous measurement control (CMC) function.
Specifications
  • Min pH (pH)-2
  • Max pH (pH)20
  • Min Temperature (° C)-5
  • Max Temperature (° C)105