Cole-Parmer LAB 865 BENCHTOP PH METER
Representative image only.
Cole-Parmer LAB 865 BENCHTOP PH METER
Cole-Parmer | Item: 1596162
₹232,000.00INR / Each
₹232,000.00INR / Each
Product Description
Includes
pH/temperature combination electrode (BlueLine 14 pH), calibration solutions, stand, USB cable, power supply, and four AA 1.5 V batteries.
Key Features
  • Rapid data transfer via USB interface
  • Manual or timer controlled data logging of measured values
  • Protocols include date, time, and ID (Lab 875P model) and sensors serial
  • numbers for GLP compliance
  • Excel Add-in included for the formatted output of all data and calibration
  • protocols
  • Output data directly to integrated printer (Lab 875P model)
More About this Item
Compact Lab 865 Precision pH Meter enables you to perform pH
measurements rapidly and reliably. It provides the maximum degree of
operating comfort, reliability and measuring certainty for all applications. The
USB interface can be used for data transmission to a PC and for software updates
of the meter. Meter features an easy-to-use graphic display with text menu for
easy handling. With the CMC (continuous measurement control) function to monitor
the congruency of measuring and calibration range for pH, you can optimize the
measuring results.