Sartorius Entris® II Toploading Balances
This innovative balance features isoCAL, LED touch technology, and 12 built-in applications
  • Handles a larger capacity
  • Hybrid screen is easy to use and improves functionality
  • Optional isoCAL internal calibration saves you time and money for accurate weighing results
  • Parts offer a high chemical resistance so there is no need to worry over spills
  • Can transfer data directly to a PC with its optional data cable – no extra software required
Sartorius Entris® II precision toploading balances offer a higher max capacity than their analytical counterparts. They are perfect for general purpose weighing. The units are constructed of chemical resistant polybutylene terephthalate, stainless steel, and glass. In addition to high capacity and durability, the balances have advanced features that include dosing, counting, percentage weighing, mixing, totalizing, animal weighing, free factor, density determination, statistics, peakhold, checkweighing, mass unit conversion, and underfloor weighing. They are up to date with a USB type C interface for connecting to your PC. They are “PC-direct”, meaning that no extra software is required. You can download your data directly to Microsoft® Word or Excel.
211,000.00 - ₹482,000.00INR / Each
31 variations of this product are available.
See All Product Options Below
31 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Power (VAC)
Availability Pricing
Sartorius BCE1202-1S Entris® II Basic Essential Toploading Balance 1200 g x 10 mg; 100 to 240 VAC, 50/60 Hz WW-11976-00 Mfr # BCE1202-1S
COMPARE
External 1200 100 to 240
₹253,000.00INR / Each
Sartorius BCE1202I-1S Entris® II Basic Essential Toploading Balance with Internal Calibration, 1200 g x 10 mg; 100 to 240 VAC, 50/60 Hz WW-11976-01 Mfr # BCE1202I-1S
COMPARE
Internal 1200 100 to 240
₹301,000.00INR / Each
Sartorius BCE2201-1S Entris® II Basic Essential Toploading Balance 2200 g x 100 mg; 100 to 240 VAC, 50/60 Hz WW-11976-04 Mfr # BCE2201-1S
COMPARE
External 2200 100 to 240
₹233,000.00INR / Each
Sartorius BCE2201I-1S Entris® II Basic Essential Toploading Balance with Internal Calibration, 2200 g x 100 mg; 100 to 240 VAC, 50/60 Hz WW-11976-05 Mfr # BCE2201I-1S
COMPARE
Internal 2200 100 to 240
₹279,000.00INR / Each
Sartorius BCE2202-1S Entris® II Basic Essential Toploading Balance 2200 g x 10 mg; 100 to 240 VAC, 50/60 Hz WW-11976-06 Mfr # BCE2202-1S
COMPARE
External 2200 100 to 240
₹273,000.00INR / Each
Sartorius BCE2202I-1S Entris® II Basic Essential Toploading Balance with Internal Calibration, 2200 g x 10 mg; 100 to 240 VAC, 50/60 Hz WW-11976-07 Mfr # BCE2202I-1S
COMPARE
Internal 2200 100 to 240
₹321,000.00INR / Each
Sartorius BCE223-1S Entris® II Basic Essential Toploading Balance 220 g x 1 mg; 100 to 240 VAC, 50/60 Hz WW-11976-08 Mfr # BCE223-1S
COMPARE
External 220 100 to 240
₹233,000.00INR / Each
Sartorius BCE223I-1S Entris® II Basic Essential Toploading Balance with Internal Calibration, 220 g x 1 mg; 100 to 240 VAC, 50/60 Hz WW-11976-09 Mfr # BCE223I-1S
COMPARE
Internal 220 100 to 240
₹279,000.00INR / Each
Sartorius BCE3202-1S Entris® II Basic Essential Toploading Balance 3200 g x 10 mg; 100 to 240 VAC, 50/60 Hz WW-11976-12 Mfr # BCE3202-1S
COMPARE
External 3200 100 to 240
₹294,000.00INR / Each
Sartorius BCE3202I-1S Entris® II Basic Essential Toploading Balance with Internal Calibration, 3200 g x 10 mg; 100 to 240 VAC, 50/60 Hz WW-11976-13 Mfr # BCE3202I-1S
COMPARE
Internal 3200 100 to 240
₹341,000.00INR / Each
Sartorius BCE323-1S Entris® II Basic Essential Toploading Balance 320 g x 1 mg; 100 to 240 VAC, 50/60 Hz WW-11976-14 Mfr # BCE323-1S
COMPARE
External 320 100 to 240
₹273,000.00INR / Each
Sartorius BCE4202-1S Entris® II Basic Essential Toploading Balance 4200 g x 10 mg; 100 to 240 VAC, 50/60 Hz WW-11976-16 Mfr # BCE4202-1S
COMPARE
External 4200 100 to 240
₹336,000.00INR / Each
Sartorius BCE4202I-1S Entris® II Basic Essential Toploading Balance with Internal Calibration, 4200 g x 10 mg; 100 to 240 VAC, 50/60 Hz WW-11976-17 Mfr # BCE4202I-1S
COMPARE
Internal 4200 100 to 240
₹381,000.00INR / Each
Sartorius BCE423-1S Entris® II Basic Essential Toploading Balance 420 g x 1 mg; 100 to 240 VAC, 50/60 Hz WW-11976-18 Mfr # BCE423-1S
COMPARE
External 420 100 to 240
₹358,000.00INR / Each
Sartorius BCE5201-1S Entris® II Basic Essential Toploading Balance 5200 g x 100 mg; 100 to 240 VAC, 50/60 Hz WW-11976-20 Mfr # BCE5201-1S
COMPARE
External 5200 100 to 240
₹253,000.00INR / Each
Sartorius BCE5201I-1S Entris® II Basic Essential Toploading Balance with Internal Calibration, 5200 g x 100 mg; 100 to 240 VAC, 50/60 Hz WW-11976-21 Mfr # BCE5201I-1S
COMPARE
Internal 5200 100 to 240
₹301,000.00INR / Each
Sartorius BCE6200-1S Entris® II Basic Essential Toploading Balance 6200 g x 1000 mg; 100 to 240 VAC, 50/60 Hz WW-11976-22 Mfr # BCE6200-1S
COMPARE
External 6200 100 to 240
₹233,000.00INR / Each
Sartorius BCE6200I-1S Entris® II Basic Essential Toploading Balance with Internal Calibration, 6200 g x 1000 mg; 100 to 240 VAC, 50/60 Hz WW-11976-23 Mfr # BCE6200I-1S
COMPARE
Internal 6200 100 to 240
₹279,000.00INR / Each
Sartorius BCE6202-1S Entris® II Basic Essential Toploading Balance 6200 g x 10 mg; 100 to 240 VAC, 50/60 Hz WW-11976-24 Mfr # BCE6202-1S
COMPARE
External 6200 100 to 240
₹442,000.00INR / Each
Sartorius BCE6202I-1S Entris® II Basic Essential Toploading Balance with Internal Calibration, 6200 g x 10 mg; 100 to 240 VAC, 50/60 Hz WW-11976-25 Mfr # BCE6202I-1S
COMPARE
Internal 6200 100 to 240
₹482,000.00INR / Each
Showing 1 - 20 of 31