Sartorius CPA Analytical and Toploading Balances
Monolithic weigh cell offers reliable precision and durability
  • Built-in, motorized calibration weight for the highest weighing accuracy
  • Electronic filters adapt to changes in ambient conditions
  • Bidirectional RS-232C interface port enables communicate with a printer or PC
Convenient, built-in, one-touch motorized calibration and automatic internal calibration adjustment (isoCAL) compensates for ambient temperature changes within the laboratory at regular intervals for greater accuracy. High-contrast, backlit display provides superior visibility under any lighting conditions.
257,000.00 - ₹404,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Sartorius CPA 225 D Balance, 100 / 220g x 0.01 Mg 1121951 Mfr # CPA 225 D
COMPARE
Internal 220
₹362,000.00INR / Each
Sartorius CPA 224 S Balance, 220g x 0.1 Mg 1121952 Mfr # CPA 224 S
COMPARE
Internal 220
₹257,000.00INR / Each
Sartorius CPA 324 S Balance, 320g x 0.1 Mg 1121953 Mfr # CPA 324 S
COMPARE
Internal 320
₹324,000.00INR / Each
Sartorius CPA 623 S Toploading Balance, 620g x 1 Mg 1121954 Mfr # CPA 623 S
COMPARE
Internal 620
₹281,000.00INR / Each
Sartorius CPA 1003 S Balance, 1, 000g x 1 Mg 1121955 Mfr # CPA 1003 S
COMPARE
Internal 1000
₹379,000.00INR / Each
Sartorius CPA 6202 S Balance, 6, 200g x 10 Mg 1121956 Mfr # CPA 6202 S
COMPARE
Internal 6200
₹306,000.00INR / Each
Sartorius CPA 16001 S Balance, 16, 000g x 100 Mg 1121957 Mfr # CPA 16001 S
COMPARE
Internal 16000
₹379,000.00INR / Each
Sartorius CPA 34001 S Balance, 34, 000g x 100 Mg 1121958 Mfr # CPA 34001 S
COMPARE
Internal 34000
₹404,000.00INR / Each
Showing 1- 8 of 8