Sartorius BT Analytical and Toploading Balances
Standard performance for all routine laboratory tasks
  • Single block aluminum alloy monolithic weighing system offers greater accuracy and durability
  • Electronic filters adapt to changing ambient temperature conditions
  • Perform a wide variety of applications with dynamic animal weighing, below-balance weighing hanger, percent weighing, counting, and mass unit conversion
Dual tare keys on left and right side of balance offer greater convenience and smooth operation in any weighing application. The easy-to-read high-contrast display provides clear viewing of results under any lighting conditions. Meet ISO and GLP compliance through PC and printer communication with the built-in RS-232 serial interface.
142,000.00 - ₹231,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Sartorius BT 124 S Balance, 120g x 0.1 Mg WW-11219-41 Mfr # BT 124 S
COMPARE
External 120
₹187,000.00INR / Each
Sartorius BT 224 S Balance, 220g x 0.1 Mg WW-11219-42 Mfr # BT 224 S
COMPARE
External 220
₹231,000.00INR / Each
Sartorius BS 223 S Balance, 220g x 1 Mg (External Calibration) WW-11219-43 Mfr # BS 223 S
COMPARE
External 220
₹142,000.00INR / Each
Sartorius BT 323 S Toploading Balance, 320g x 1 Mg WW-11219-44 Mfr # BT 323 S
COMPARE
External 320
₹190,000.00INR / Each
Sartorius BT 423 S Toploading Balance, 420g x 1 Mg WW-11219-45 Mfr # BT 423 S
COMPARE
External 420
₹207,000.00INR / Each
Sartorius BT 2202 S Balance, 2, 200g x 10 Mg WW-11219-46 Mfr # BT 2202 S
COMPARE
External 2200
₹172,000.00INR / Each
Sartorius BT 4202 S Balance, 4, 200g x 10 Mg WW-11219-47 Mfr # BT 4202 S
COMPARE
External 4200
₹190,000.00INR / Each
Showing 1- 7 of 7