Saint-Gobain Single-Use 3D Bioprocess Bags for Cylindrical Tanks with 3 Ports and 30"L Tubing
Multilayer bags provide outstanding bio and chemical compatibility, structural strength, and gas barrier—excellent for many demanding bioprocess applications
  • Easily integrate into your single-use process applications
  • Minimize the risk of cross-contamination and reduce cleaning time
  • 3D bags are often cuboidal in shape—perfect for use in tanks or containers
  • Three ports with C-Flex® 374 tubing and fittings, including a needle-free sampling port
  • Free of any animal-derived components for greater process purity
  • Manufactured and packed in an ISO Class 7 certified cleanroom
  • QSR and ISO 9001:2015 compliant
  • Meet the requirements of USP Class VI and ISO 10993-3 testing
These single-use bioprocess bags feature multilayer films in different materials to provide the best biocompatibility, chemical resistance, and gas barrier to meet your bioprocessing applications. The LLDPE fluid contact layer offers outstanding biocompatibility with broad chemical compatibility for safe handling of a wide range of biopharmaceutical liquids. The EVOH middle layer ensures minimum gas permeation, ensuring the integrity of the fluid. The impact and tear resistant outside nylon layer offers outstanding durability. A multi-material tie-structure bonds the three layers, adding to the overall performance of the bag. Bags are ideal for use in buffer and media storage, bioreactor feed and harvest, and chromatography feed.
A 3D bag consists of multilayer sheets sealed together to often form a cuboidal shape and fits inside a supporting tank or container. These collapsible bags minimize the risk of cross-contamination and reduce cleaning and turnaround time. Each bag features three ports with 30” long clear C-Flex® 374 tubing, MPX inserts (on two ports) or female locking Luer with needle-free sampling port, and pinch clamps. Use bags in temperatures ranging from –80 to 60°C (–112 to 140°F). Bags are sterilized by gamma irradiation, ports are capped and covered, and systems are double bagged for integrity.
356,000.00 - ₹803,000.00INR / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters)
Availability Pricing
Saint Gobain Single-Use 3D Bioprocess Bags for Cylindrical Tanks, 50 L, 3 Ports, 30"L Tubing; 10/Pk 4852415 Mfr # BP0050LD
COMPARE
50
₹663,000.00INR / Pkg of 10
Saint Gobain Single-Use 3D Bioprocess Bags for Cylindrical Tanks, 100 L, 3 Ports, 30"L Tubing; 10/Pk 4852416 Mfr # BP0100LD
COMPARE
100
₹730,000.00INR / Pkg of 10
Saint Gobain Single-Use 3D Bioprocess Bags for Cylindrical Tanks, 200 L, 3 Ports, 30"L Tubing; 10/Pk 4852417 Mfr # BP0200LD
COMPARE
200
₹803,000.00INR / Pkg of 10
Saint Gobain Single-Use 3D Bioprocess Bags for Cylindrical Tanks, 300 L, 3 Ports, 30"L Tubing; 4/Pk 4852418 Mfr # BP0300LD
COMPARE
300
₹356,000.00INR / Pkg of 4
Saint Gobain Single-Use 3D Bioprocess Bags for Cylindrical Tanks, 500 L, 3 Ports, 30"L Tubing; 4/Pk 4852419 Mfr # BP0500LD
COMPARE
500
₹388,000.00INR / Pkg of 4