Saint Gobain Single-Use 3D Bioprocess Bags for Cylindrical Tanks, 500 L, 3 Ports, 30"L Tubing; 4/Pk

Representative image only.

Saint Gobain Single-Use 3D Bioprocess Bags for Cylindrical Tanks, 500 L, 3 Ports, 30"L Tubing; 4/Pk

Saint Gobain | Mfr: BP0500LD| Item: 4852419

Multilayer bags provide outstanding bio and chemical compatibility, structural strength, and gas barrier—excellent for many demanding bioprocess applications

₹388,000.00INR / Pkg of 4

₹388,000.00INR / Pkg of 4
Product Description

Includes

Ports 1 and 2: 30" long clear C-Flex® 374 tubing (1/2"ID x 3/4"OD), MPX insert, MPX cap, and pinch clamp.
Port 3: 30" long clear C-Flex tubing (1/8"ID x 1/4"OD), female locking Luer fitting with needle-free sampling port, Luer cap, and pinch clamp.

Key Features

  • Easily integrate into your single-use process applications
  • Minimize the risk of cross-contamination and reduce cleaning time
  • 3D bags are often cuboidal in shape—perfect for use in tanks or containers
  • Three ports with C-Flex® 374 tubing and fittings, including a needle-free sampling port
  • Free of any animal-derived components for greater process purity
  • Manufactured and packed in an ISO Class 7 certified cleanroom
  • QSR and ISO 9001:2015 compliant
  • Meet the requirements of USP Class VI and ISO 10993-3 testing
  • 500 L apaciry

More About this Item

These single-use bioprocess bags feature multilayer films in different materials to provide the best biocompatibility, chemical resistance, and gas barrier to meet your bioprocessing applications. The LLDPE fluid contact layer offers outstanding biocompatibility with broad chemical compatibility for safe handling of a wide range of biopharmaceutical liquids. The EVOH middle layer ensures minimum gas permeation, ensuring the integrity of the fluid. The impact and tear resistant outside nylon layer offers outstanding durability. A multi-material tie-structure bonds the three layers, adding to the overall performance of the bag. Bags are ideal for use in buffer and media storage, bioreactor feed and harvest, and chromatography feed.

A 3D bag consists of multilayer sheets sealed together to often form a cuboidal shape and fits inside a supporting tank or container. These collapsible bags minimize the risk of cross-contamination and reduce cleaning and turnaround time. Each bag features three ports with 30” long clear C-Flex® 374 tubing, MPX inserts (on two ports) or female locking Luer with needle-free sampling port, and pinch clamps. Use bags in temperatures ranging from –80 to 60°C (–112 to 140°F). Bags are sterilized by gamma irradiation, ports are capped and covered, and systems are double bagged for integrity.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Specifications
  • ColorTranslucent
  • Capacity (Liters)500
  • MaterialLLDPE (inside), EVOH (middle), nylon (outside), and multiple materials (tie structure)
  • Length (in)15
  • Width (in)15
  • Height (InchShort)30.5
  • Length (cm)38.1
  • Width (cm)38.1
  • Height (cm)77.47
  • Thickness (mil)12
  • DescriptionSingle-Use 3D Bioprocess Bags for Cylindrical Tanks, 500 L, 3 Ports, 30"L Tubing; 4/Pk
Reviews
Q&A