Rite in the Rain All-Weather Spiral Notebooks

Ensure your data survives weather conditions
22,900.00 - ₹45,900.00INR / Pkg of 12

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Color Number of Pages Description
Availability Pricing
Rite in the Rain 393N-6 4-5/8" x 7" spiral notebook - horizontal line pattern 0657008 Mfr # 393
COMPARE
Yellow 64 4-5/8" x 7" spiral notebook - horizontal line pattern
₹45,900.00INR / Pkg of 12
Rite in the Rain 135-12 Spiral 3 x 5" Pocket Notebook, Lined Paper; 12/Pk 0657010 Mfr # 135
COMPARE
Yellow 50 Spiral 3 x 5" Pocket Notebook, Lined Paper; 12/Pk
₹22,900.00INR / Pkg of 12
Rite in the Rain 146-12 Spiral 4 x 6" Pocket Notebook, Lined Paper; 12/Pk 0657012 Mfr # 146
COMPARE
Yellow 50 Spiral 4 x 6" Pocket Notebook, Lined Paper; 12/Pk
₹32,000.00INR / Pkg of 12
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat