ResinTech Reverse Osmosis (RO) Water Purification System Accessories
Keep your reverse osmosis pretreatment system producing high-quality water
  • Quick-change cartridges simplify system maintenance
  • Wall-mounted design saves on limited bench space
  • Made in the USA!
These replacement accessories are compatible with the ResinTech HPL-RO reverse osmosis pretreatment system, model 99285-51. Pretreatment system sold separately.
35,500.00 - ₹52,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Compatible with Media
Availability Pricing
ResinTech AFK-005 HPL-RO Pretreatment Cartridge Kit (5.0/GAC / 1.0) 9928552 Mfr # AFK-005
COMPARE
Replacement Pretreatment Kit 99285-51 GAC carbon filter
₹35,500.00INR / Each
ResinTech AM-12-7010 RO Membrane - 100 GPD 9928553 Mfr # AM-12-7010
COMPARE
Membrane, 100 GPD 99285-51 Polyamide membrane
₹52,000.00INR / Each