ResinTech AFK-005 HPL-RO Pretreatment Cartridge Kit (5.0/GAC / 1.0)
Representative image only.
ResinTech AFK-005 HPL-RO Pretreatment Cartridge Kit (5.0/GAC / 1.0)
ResinTech | Mfr: AFK-005| Item: 9928552
Keep your reverse osmosis pretreatment system producing high-quality water
₹35,500.00INR / Each
₹35,500.00INR / Each
Product Description
Includes
5.0 micron prefilter, GAC carbon filter, and 1.0 micron post filter.
Key Features
  • Quick-change cartridges simplify system maintenance
  • Wall-mounted design saves on limited bench space
  • Made in the USA!
  • 1 year warranty
More About this Item
These replacement accessories are compatible with the ResinTech HPL-RO reverse osmosis pretreatment system, model 99285-51. Pretreatment system sold separately.
Replacement cartridge kit for Aries HPL-RO reverse osmosis pretreatment system, model 99285-51. Pretreatment system sold separately.
1 Year Warranty
Specifications
  • MediaGAC carbon filter
  • Pore size (µm)1
  • Compatible with99285-51
  • ApplicationsAutoclave, glassware washing and rinsing, general lab equiptment, Pre-treated water for Type I systems
  • DescriptionReplacement Pretreatment Kit