ResinTech AM-12-7010 RO Membrane - 100 GPD
Representative image only.
ResinTech AM-12-7010 RO Membrane - 100 GPD
ResinTech | Mfr: AM-12-7010| Item: 9928553
Keep your reverse osmosis pretreatment system producing high-quality water
₹52,000.00INR / Each
₹52,000.00INR / Each
Product Description
Key Features
  • Quick-change cartridges simplify system maintenance
  • Wall-mounted design saves on limited bench space
  • Made in the USA!
  • Thin film composite PA membrane with 100 GPD output
  • 1 year warranty
More About this Item
These replacement accessories are compatible with the ResinTech HPL-RO reverse osmosis pretreatment system, model 99285-51. Pretreatment system sold separately.
Replacement cartridge kit for Aries HPL-RO reverse osmosis pretreatment system, model 99285-51. Pretreatment system sold separately.
1 Year Warranty
Specifications
  • MediaPolyamide membrane
  • Compatible with99285-51
  • ApplicationsAutoclave, glassware washing and rinsing, general lab equiptment, Pre-treated water for Type I systems
  • Max Flow Rate (LPH)15.8
  • Max Flow Rate (GPH)4.17
  • DescriptionMembrane, 100 GPD