Qsonica Q55 Sonicator Ultrasonic Processor
Compact and economical ultrasonic processor occupying less bench space than any unit on the market
  • Smallest unit available
  • Thumb-switch or continuous operation
  • Simple and effective operation
This model is effective for standard cell disruption and many other small volume applications. Probes are available in three different sizes. Order probes and accessories separately.
474,000.00 - ₹607,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Watts
Availability Pricing
Qsonica Q55-110 Q55 Sonicator Ultrasonic Processor; 110 VAC 0471251 Mfr # Q55-110
COMPARE
60 110 55
₹607,000.00INR / Each
Qsonica Q55-220 Q55 Sonicator Ultrasonic Processor; 220 VAC 0471252 Mfr # Q55-220
COMPARE
50 220 55
₹474,000.00INR / Each