QLA Vessels Compatible with Agilent V-Series Dissolution Apparatus
TruCenter Vessels offer superior centering and vertical verticality for V-series baths
  • Turned on a lathe during manufacturing to find the exact center Unique serial number is acid etched on each vessel 1000 mL capacity
Glass vessels offer good thermal conductivity and are highly resistance to scratching. Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Select PEAK vessels with a unique cone shape molded into the bottom to improve hydrodynamics, preventing cone-shaped formations of material at the base of the vessel under the paddle.
76,100.00 - ₹129,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Bottle Color Material Special Features
Availability Pricing
QLA Standard Vessel for V-Series, TruCenter, No Collar, 1000 mL, Amber Glass; Serialized 4130630 Mfr # GLA900-AVK
COMPARE
Amber Glass Standard
₹107,000.00INR / Each
QLA Standard Vessel for V-Series, TruCenter, No Collar, 1000 mL, Clear Glass; Serialized 4130641 Mfr # GLA900-VK
COMPARE
Clear Glass Standard
₹76,100.00INR / Each
QLA PEAK Vessel for V-Series, TruCenter, No Collar, 1000 mL, Clear Glass; Serialized 4130656 Mfr # GLA9PK-VK
COMPARE
Clear Glass PEAK style
₹129,000.00INR / Each