QLA PEAK Vessel for V-Series, TruCenter, No Collar, 1000 mL, Clear Glass; Serialized
Representative image only.
QLA PEAK Vessel for V-Series, TruCenter, No Collar, 1000 mL, Clear Glass; Serialized
| Mfr: GLA9PK-VK | Item: 4130656
TruCenter Vessels offer superior centering and vertical verticality for V-series baths
₹129,000.00INR / Each
₹129,000.00INR / Each
Product Description
Key Features
  • Turned on a lathe during manufacturing to find the exact center Unique serial number is acid etched on each vessel 1000 mL capacity
  • PEAK-style vessel
  • Clear glass
More About this Item
Glass vessels offer good thermal conductivity and are highly resistance to scratching. Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Select PEAK vessels with a unique cone shape molded into the bottom to improve hydrodynamics, preventing cone-shaped formations of material at the base of the vessel under the paddle.
.
PEAK vessels do not comply with USP specifications and cannot be substituted in existing applications.
Specifications
  • Capacity (mL)1000
  • MaterialGlass
  • Bottle ColorClear
  • Special FeaturesPEAK style
  • DescriptionPEAK Vessel for V-Series, TruCenter, No Collar, 1000 mL, Clear Glass; Serialized