QLA Standard Vessel for V-Series, TruCenter, No Collar, 1000 mL, Amber Glass; Serialized
Representative image only.
QLA Standard Vessel for V-Series, TruCenter, No Collar, 1000 mL, Amber Glass; Serialized
| Mfr: GLA900-AVK| Item: 4130630
TruCenter Vessels offer superior centering and vertical verticality for V-series baths
₹107,000.00INR / Each
₹107,000.00INR / Each
Product Description
Key Features
  • Turned on a lathe during manufacturing to find the exact center Unique serial number is acid etched on each vessel 1000 mL capacity
  • Standard vessel
  • Amber glass is ideal for light-sensitive compounds
More About this Item
Glass vessels offer good thermal conductivity and are highly resistance to scratching. Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Select PEAK vessels with a unique cone shape molded into the bottom to improve hydrodynamics, preventing cone-shaped formations of material at the base of the vessel under the paddle.
.
Specifications
  • Capacity (mL)1000
  • MaterialGlass
  • Bottle ColorAmber
  • Special FeaturesStandard
  • DescriptionStandard Vessel for V-Series, TruCenter, No Collar, 1000 mL, Amber Glass; Serialized