QLA Sotax-Compatible Sampling Cannula
Consistent sampling with QLA Sotax-compatible Cannula
  • Matching cannula to stopper ensures greater sampling position consistency Made of stainless steel that will not absorb or interfere with active ingredients
For dissolution tests, taking reliable samples is critical. QLA probes, cannula, and stoppers assure consistent sampling position within the mixing vessel, which means more accurate testing.
34,800.00 - ₹64,200.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (") Material Description
Availability Pricing
QLA Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; Thru-Head Mount 4130212 Mfr # CAN013-ST
COMPARE
13 316 stainless steel Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; Thru-Head Mount
₹34,800.00INR / Each
QLA Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; Preset with Stopper for 500 mL 4130213 Mfr # CAN013-ST-5
COMPARE
13 316 stainless steel Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; Preset with Stopper for 500 mL
₹55,500.00INR / Each
QLA Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; 2-Position Stopper for 500 & 900 mL 4130214 Mfr # CAN013-ST-5-9
COMPARE
13 316 stainless steel Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; 2-Position Stopper for 500 & 900 mL
₹64,200.00INR / Each
QLA Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; Preset with Stopper for 900 mL 4130215 Mfr # CAN013-ST-9
COMPARE
13 316 stainless steel Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; Preset with Stopper for 900 mL
₹55,500.00INR / Each
QLA Bent 316 SS Cannula for Sotax, 15"L, SS Luer Lock; Preset with Stopper for 900 mL 4130219 Mfr # CAN015-ST-9
COMPARE
15 316 stainless steel Bent 316 SS Cannula for Sotax, 15"L, SS Luer Lock; Preset with Stopper for 900 mL
₹58,900.00INR / Each