QLA Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; Thru-Head Mount
Representative image only.
QLA Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; Thru-Head Mount
| Mfr: CAN013-ST| Item: 4130212
Consistent sampling with QLA Sotax-compatible Cannula
₹34,800.00INR / Each
₹34,800.00INR / Each
Buy It Together
Current Item
QLA Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; Thru-Head Mount
QLA Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; Thru-Head Mount
4130212
₹34,800.00INR / Each
+
QLA Adjustable Cannula Stopper for Sotax
4130281
₹18,500.00INR / Each
=
Total Price 53300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency Made of stainless steel that will not absorb or interfere with active ingredients
  • Thru-head mount
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannula, and stoppers assure consistent sampling position within the mixing vessel, which means more accurate testing.
.
Specifications
  • Material316 stainless steel
  • Length (")13
  • DescriptionBent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; Thru-Head Mount