QLA Adjustable Cannula Stopper for Sotax
Representative image only.
QLA Adjustable Cannula Stopper for Sotax
| Mfr: CANSTP-ST| Item: 4130281
QLA accessories help provide more precise and consistent dissolution results
₹18,500.00INR / Each
₹18,500.00INR / Each
Product Description
Key Features
  • Use with 41302-12 cannula
More About this Item
Use stopper with cannula to ensure the sample is always taken from the same depth.
.
Specifications
  • Compatible WithSotax
  • DescriptionAdjustable Cannula Stopper for Sotax