QLA Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; 2-Position Stopper for 500 & 900 mL

Representative image only.

QLA Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; 2-Position Stopper for 500 & 900 mL

| Mfr: CAN013-ST-5-9| Item: 4130214

Consistent sampling with QLA Sotax-compatible Cannula

₹64,200.00INR / Each

₹64,200.00INR / Each

Buy It Together

Current Item

QLA Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; 2-Position Stopper for 500 & 900 mL
QLA Bent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; 2-Position Stopper for 500 & 900 mL
4130214

₹64,200.00INR / Each
+
QLA Adjustable Cannula Stopper for Sotax
4130281

₹18,500.00INR / Each
=
Total Price 82700.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Matching cannula to stopper ensures greater sampling position consistency Made of stainless steel that will not absorb or interfere with active ingredients
  • Adjustable, 2-position stopper for 500 or 900 mL vessel

More About this Item

For dissolution tests, taking reliable samples is critical. QLA probes, cannula, and stoppers assure consistent sampling position within the mixing vessel, which means more accurate testing.

.

Specifications
  • Material316 stainless steel
  • Length (")13
  • DescriptionBent 316 SS Cannula for Sotax, 13"L, SS Luer Lock; 2-Position Stopper for 500 & 900 mL
Reviews
Q&A