QLA Pharmatest-Compatible Baskets
QLA Baskets for dissolution testing with your Pharmatest apparatus
  • Made of 316 electropolished stainless steel All baskets and shafts are laser marked with unique serial numbers
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Pharmatest test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
45,000.00 - ₹175,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Mesh Size Description
Availability Pricing
QLA Clip Style Basket for Pharmatest, 10 Mesh, 316 SS; Serialized 4130133 Mfr # BSK010-PT
COMPARE
316 stainless steel 10 Clip Style Basket for Pharmatest, 10 Mesh, 316 SS; Serialized
₹45,200.00INR / Each
QLA Clip Style Basket for Pharmatest, 20 Mesh, 316 SS; Serialized 4130141 Mfr # BSK020-PT
COMPARE
316 stainless steel 20 Clip Style Basket for Pharmatest, 20 Mesh, 316 SS; Serialized
₹46,100.00INR / Each
QLA Clip Style Basket for Pharmatest, 40 Mesh USP, 316 SS; Serialized 4130151 Mfr # BSK040-PT
COMPARE
316 stainless steel 40 Clip Style Basket for Pharmatest, 40 Mesh USP, 316 SS; Serialized
₹45,000.00INR / Each
QLA Clip Style Basket for Pharmatest, 40 Mesh, PTFE Coated over 316 SS; Serialized 4130173 Mfr # BSKPTP-40
COMPARE
PTFE-coated 316 SS 40 Clip Style Basket for Pharmatest, 40 Mesh, PTFE Coated over 316 SS; Serialized
₹83,900.00INR / Each
QLA Suppository Basket for Pharmatest; Plastic 4130197 Mfr # BSKSUP-PT
COMPARE
Plastic Suppository Basket for Pharmatest; Plastic
₹55,500.00INR / Each
QLA Clip Style Basket for Pharmatest, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130844 Mfr # CUSBSK-20/100PT
COMPARE
316 stainless steel 100 (plus 20 mesh stability lining) Clip Style Basket for Pharmatest, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
₹175,000.00INR / Each
QLA Clip Style Basket for Pharmatest, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130851 Mfr # CUSBSK-20/150PT
COMPARE
316 stainless steel 150 (plus 20 mesh stability lining) Clip Style Basket for Pharmatest, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
₹175,000.00INR / Each
Showing 1- 7 of 7