QLA Clip Style Basket for Pharmatest, 40 Mesh, PTFE Coated over 316 SS; Serialized
Representative image only.
QLA Clip Style Basket for Pharmatest, 40 Mesh, PTFE Coated over 316 SS; Serialized
| Mfr: BSKPTP-40| Item: 4130173
QLA Baskets for dissolution testing with your Pharmatest apparatus
₹83,900.00INR / Each
₹83,900.00INR / Each
Buy It Together
Current Item
QLA Clip Style Basket for Pharmatest, 40 Mesh, PTFE Coated over 316 SS; Serialized
QLA Clip Style Basket for Pharmatest, 40 Mesh, PTFE Coated over 316 SS; Serialized
4130173
₹83,900.00INR / Each
+
QLA Basket Holder, Universal Compatibility; 6 Position
4130170
₹39,100.00INR / Each
=
Total Price 123000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of 316 electropolished stainless steel All baskets and shafts are laser marked with unique serial numbers
  • 40 mesh
  • PTFE-coated 316 SS
More About this Item
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Pharmatest test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • MaterialPTFE-coated 316 SS
  • Mesh Size40
  • DescriptionClip Style Basket for Pharmatest, 40 Mesh, PTFE Coated over 316 SS; Serialized