QLA Clip Style Basket for Pharmatest, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
Representative image only.
QLA Clip Style Basket for Pharmatest, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
| Mfr: CUSBSK-20/150PT| Item: 4130851
QLA Baskets for dissolution testing with your Pharmatest apparatus
₹175,000.00INR / Each
₹175,000.00INR / Each
Buy It Together
Current Item
QLA Clip Style Basket for Pharmatest, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
QLA Clip Style Basket for Pharmatest, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
4130851
₹175,000.00INR / Each
+
QLA Basket Holder, Universal Compatibility; 6 Position
4130170
₹39,100.00INR / Each
=
Total Price 214100.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of 316 electropolished stainless steel All baskets and shafts are laser marked with unique serial numbers
  • 150 mesh
More About this Item
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Pharmatest test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • Material316 stainless steel
  • Mesh Size150 (plus 20 mesh stability lining)
  • DescriptionClip Style Basket for Pharmatest, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized