QLA Multibrand-Compatible Cannula
Achieve consistent sampling using QLA cannulas
  • Matching cannula to stopper ensures greater sampling position consistency
For dissolution tests, taking reliable samples is critical. QLA cannulas and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
30,600.00 - ₹35,600.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (") Material Description
Availability Pricing
QLA Bent PEEK Cannula for Erweka/Hanson, 15"L, 1/8" OD; PEEK Luer Lock 4130217 Mfr # CAN015-PH
COMPARE
15 PEEK Bent PEEK Cannula for Hanson, 15"L, 1/8" OD; PEEK Luer Lock
₹31,500.00INR / Each
QLA Bent 316 SS Cannula for Erweka/Hanson, 15"L, 1/8" OD; SS Luer Lock 4130218 Mfr # CAN015-SH
COMPARE
15 316 stainless steel Bent 316 SS Cannula for Hanson, 15"L, 1/8" OD; SS Luer Lock
₹35,600.00INR / Each
QLA Bent PEEK Cannula for Erweka/Hanson/Pharmatest, 4.75"L, 1/8" OD; PEEK Luer Lock 4130231 Mfr # CAN475-PH
COMPARE
4 3/4 PEEK Bent PEEK Cannula for Hanson, 4.75"L, 1/8" OD; PEEK Luer Lock
₹30,600.00INR / Each
QLA Bent 316 SS Cannula for Erweka/Hanson/Pharmatest, 4.75"L, 1/8" OD; SS luer Lock 4130233 Mfr # CAN475-SH
COMPARE
4 3/4 316 stainless steel Bent 316 SS Cannula for Hanson, 4.75"L, 1/8" OD; SS luer Lock
₹30,600.00INR / Each
QLA Bent PEEK Cannula for Erweka/Hanson/Pharmatest, 7.75"L, 1/8" OD; PEEK Luer Lock 4130249 Mfr # CAN775-PH
COMPARE
7 3/4 PEEK Bent PEEK Cannula for Hanson, 7.75"L, 1/8" OD; PEEK Luer Lock
₹32,600.00INR / Each
QLA Bent 316 SS Cannula for Erweka/Hanson/Pharmatest, 7.75"L, 1/8" OD; SS Luer Lock 4130250 Mfr # CAN775-SH
COMPARE
7 3/4 316 stainless steel Bent 316 SS Cannula for Hanson, 7.75"L, 1/8" OD; SS Luer Lock
₹33,500.00INR / Each