QLA Hanson-Compatible Baskets
QLA Baskets for dissolution testing with your Hanson apparatus
  • Made of 316 electropolished stainless steel All baskets are laser marked with unique serial numbers
These QLA baskets and shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Hanson test apparatus. All (except plastic suppository baskets) are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
39,500.00 - ₹175,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Mesh Size Description
Availability Pricing
QLA Clip Style Basket for Hanson, 10 Mesh, 316 SS; Serialized 4130132 Mfr # BSK010-HR
COMPARE
316 stainless steel 10 QLA Clip Style Basket for Hanson, 10 Mesh, 316 SS; Serialized
₹45,200.00INR / Each
QLA Clip Style Basket for Hanson, 20 Mesh, 316 SS; Serialized 4130140 Mfr # BSK020-HR
COMPARE
316 stainless steel 20 QLA Clip Style Basket for Hanson, 20 Mesh, 316 SS; Serialized
₹46,100.00INR / Each
QLA Clip Style Basket for Hanson, 40 Mesh USP, 316 SS; Serialized 4130150 Mfr # BSK040-HRP
COMPARE
316 stainless steel 40 QLA Clip Style Basket for Hanson, 40 Mesh USP, 316 SS; Serialized
₹39,500.00INR / Each
QLA Clip Style Basket for Hanson, 100 Mesh, 316 SS; Serialized 4130161 Mfr # BSK100-HR
COMPARE
316 stainless steel 100 QLA Clip Style Basket for Hanson, 100 Mesh, 316 SS; Serialized
₹82,500.00INR / Each
QLA Clip Style Basket for Hanson, 140 Mesh, 316 SS; Serialized 4130164 Mfr # BSK140-HR
COMPARE
316 stainless steel 140 QLA Clip Style Basket for Hanson, 140 Mesh, 316 SS; Serialized
₹123,000.00INR / Each
QLA Clip Style Basket for Hanson, 40 Mesh, PTFE Coated 316 SS; Serialized 4130172 Mfr # BSKPTH-40
COMPARE
PTFE-coated 316 SS 40 QLA Clip Style Basket for Hanson, 40 Mesh, PTFE Coated 316 SS; Serialized
₹81,800.00INR / Each
QLA Suppository Basket for Hanson; Plastic 4130196 Mfr # BSKSUP-HR
COMPARE
Plastic QLA Suppository Basket for Hanson; Plastic
₹55,500.00INR / Each
QLA Clip Style Basket for Hanson, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130843 Mfr # CUSBSK-20/100HR
COMPARE
316 stainless steel 100 (plus 20 mesh stability lining) QLA Clip Style Basket for Hanson, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
₹175,000.00INR / Each
QLA Clip Style Basket for Hanson, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130850 Mfr # CUSBSK-20/150HR
COMPARE
316 stainless steel 150 (plus 20 mesh stability lining) QLA Clip Style Basket for Hanson, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
₹175,000.00INR / Each
Showing 1- 9 of 9