QLA Clip Style Basket for Hanson, 20 Mesh, 316 SS; Serialized
Representative image only.
QLA Clip Style Basket for Hanson, 20 Mesh, 316 SS; Serialized
| Mfr: BSK020-HR| Item: 4130140
QLA Baskets for dissolution testing with your Hanson apparatus
₹46,100.00INR / Each
₹46,100.00INR / Each
Buy It Together
Current Item
QLA Clip Style Basket for Hanson, 20 Mesh, 316 SS; Serialized
QLA Clip Style Basket for Hanson, 20 Mesh, 316 SS; Serialized
4130140
₹46,100.00INR / Each
+
QLA Basket Holder, Universal Compatibility; 12 Position
4130169
₹49,600.00INR / Each
=
Total Price 95700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of 316 electropolished stainless steel All baskets are laser marked with unique serial numbers
  • 20 mesh
More About this Item
These QLA baskets and shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Hanson test apparatus. All (except plastic suppository baskets) are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • Material316 stainless steel
  • Mesh Size20
  • DescriptionQLA Clip Style Basket for Hanson, 20 Mesh, 316 SS; Serialized