QLA Suppository Basket for Hanson; Plastic
Representative image only.
QLA Suppository Basket for Hanson; Plastic
| Mfr: BSKSUP-HR| Item: 4130196
QLA Baskets for dissolution testing with your Hanson apparatus
₹55,500.00INR / Each
₹55,500.00INR / Each
Buy It Together
Current Item
QLA Suppository Basket for Hanson; Plastic
QLA Suppository Basket for Hanson; Plastic
4130196
₹55,500.00INR / Each
+
QLA Basket Holder, Universal Compatibility; 12 Position
4130169
₹49,600.00INR / Each
=
Total Price 105100.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of 316 electropolished stainless steel All baskets are laser marked with unique serial numbers
  • Suppository basket
More About this Item
These QLA baskets and shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Hanson test apparatus. All (except plastic suppository baskets) are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • MaterialPlastic
  • DescriptionQLA Suppository Basket for Hanson; Plastic