QLA Dissolution Apparatus Paddles
Finely tooled dissolution paddles for consistent spin and less wobble
  • Made of electropolished 316 stainless steel Rotate smoothly, without significant wobble All paddles are laser-etched with individual serial numbers
Each paddle rotates smoothly and consistently, without wobble, meeting USP specifications. The 316 stainless steel shafts are strong and highly resistant to bending, while the electropolished surface removes surface impurities and repels corrosion.
53,000.00 - ₹91,400.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Length (InchShort) Compatible With Description
Availability Pricing
QLA Paddle Compatible with Agilent V-Series Dissolution Apparatus, 14.5"; Electropolished 316 SS 4130867 Mfr # EPD013-01
COMPARE
14 1/2 Agilent V-series Paddle for Agilent V-Series, 14.5"; Electropolished 316 SS
₹55,100.00INR / Each
QLA Paddle for VanKel/Varian, 15"; Electropolished 316 SS 4130868 Mfr # EPD015-01
COMPARE
15 VanKel/Varian Paddle for VanKel/Varian, 15"; Electropolished 316 SS
₹53,000.00INR / Each
QLA Paddle for Hanson SR8-Plus, 16"; Electropolished 316 SS 4130869 Mfr # EPD016-01
COMPARE
16 Hanson SR8-Plus Paddle for Hanson SR8-Plus, 16"; Electropolished 316 SS
₹64,700.00INR / Each
QLA Paddle for Distek 2100, 16.5"; Electropolished 316 SS 4130870 Mfr # EPD016-02
COMPARE
16 1/2 Distek 2100 Paddle for Distek 2100, 16.5"; Electropolished 316 SS
₹64,700.00INR / Each
QLA Paddle for VanKel/Varian, 19"; Electropolished 316 SS 4130871 Mfr # EPD019-01
COMPARE
19 VanKel/Varian Paddle for VanKel/Varian, 19"; Electropolished 316 SS
₹68,600.00INR / Each
QLA Paddle Compatible with Agilent Dissolution Apparatus, 21"; Electropolished 316 SS 4130872 Mfr # EPD021-01
COMPARE
21 Agilent Paddle for Agilent, 21"; Electropolished 316 SS
₹80,200.00INR / Each
QLA Paddle Compatible with Agilent Dissolution Apparatus, 24"; Electropolished 316 SS 4130873 Mfr # EPD024-01
COMPARE
24 Agilent Paddle for Agilent, 24"; Electropolished 316 SS
₹91,400.00INR / Each
Showing 1- 7 of 7