QLA Paddle Compatible with Agilent Dissolution Apparatus, 21"; Electropolished 316 SS
Representative image only.
QLA Paddle Compatible with Agilent Dissolution Apparatus, 21"; Electropolished 316 SS
| Mfr: EPD021-01 | Item: 4130872
Finely tooled dissolution paddles for consistent spin and less wobble
₹80,200.00INR / Each
₹80,200.00INR / Each
Product Description
Key Features
  • Made of electropolished 316 stainless steel Rotate smoothly, without significant wobble All paddles are laser-etched with individual serial numbers
  • For Agilent apparatus
  • 21"L
More About this Item
Each paddle rotates smoothly and consistently, without wobble, meeting USP specifications. The 316 stainless steel shafts are strong and highly resistant to bending, while the electropolished surface removes surface impurities and repels corrosion.
.
Specifications
  • MaterialElectropolished 316 SS
  • Compatible WithAgilent
  • Length (InchShort)21
  • Length (cm)53.3
  • DescriptionPaddle for Agilent, 21"; Electropolished 316 SS