QLA Paddle for Hanson SR8-Plus, 16"; Electropolished 316 SS

Representative image only.

QLA Paddle for Hanson SR8-Plus, 16"; Electropolished 316 SS

| Mfr: EPD016-01 | Item: 4130869

Finely tooled dissolution paddles for consistent spin and less wobble

₹64,700.00INR / Each

₹64,700.00INR / Each
Product Description

Key Features

  • Made of electropolished 316 stainless steel Rotate smoothly, without significant wobble All paddles are laser-etched with individual serial numbers
  • For Hanson SR8-Plus apparatus

More About this Item

Each paddle rotates smoothly and consistently, without wobble, meeting USP specifications. The 316 stainless steel shafts are strong and highly resistant to bending, while the electropolished surface removes surface impurities and repels corrosion.

.

Specifications
  • MaterialElectropolished 316 SS
  • Compatible WithHanson SR8-Plus
  • Length (InchShort)16
  • Length (cm)40.6
  • DescriptionPaddle for Hanson SR8-Plus, 16"; Electropolished 316 SS
Reviews
Q&A