QLA Caleva-Compatible Baskets
QLA Baskets for dissolution testing with your Caleva apparatus
  • Made of 316 electropolished stainless steel All baskets are laser marked with unique serial numbers
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Caleva test apparatus. All (except plastic suppository baskets) are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
47,700.00 - ₹123,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Mesh Size Description
Availability Pricing
QLA Clip Style Basket for Caleva, 10 Mesh, 316 SS; Serialized 4130128 Mfr # BSK010-CA
COMPARE
316 stainless steel 10 Clip Style Basket for Caleva, 10 Mesh, 316 SS; Serialized
₹48,500.00INR / Each
QLA Clip Style Basket for Caleva, 20 Mesh, 316 SS; Serialized 4130136 Mfr # BSK020-CA
COMPARE
316 stainless steel 20 Clip Style Basket for Caleva, 20 Mesh, 316 SS; Serialized
₹54,000.00INR / Each
QLA Clip Style Basket for Caleva, 40 Mesh USP, 316 SS; Serialized 4130146 Mfr # BSK040-CA
COMPARE
316 stainless steel 40 Clip Style Basket for Caleva, 40 Mesh USP, 316 SS; Serialized
₹47,700.00INR / Each
QLA Clip Style Basket for Caleva, 140 Mesh, 316 SS; Serialized 4130163 Mfr # BSK140-CA
COMPARE
316 stainless steel 140 Clip Style Basket for Caleva, 140 Mesh, 316 SS; Serialized
₹123,000.00INR / Each
QLA Suppository Basket for Caleva; Plastic 4130193 Mfr # BSKSUP-CA
COMPARE
Plastic Suppository Basket for Caleva; Plastic
₹55,500.00INR / Each