QLA Clip Style Basket for Caleva, 10 Mesh, 316 SS; Serialized
Representative image only.
QLA Clip Style Basket for Caleva, 10 Mesh, 316 SS; Serialized
| Mfr: BSK010-CA| Item: 4130128
QLA Baskets for dissolution testing with your Caleva apparatus
₹48,500.00INR / Each
₹48,500.00INR / Each
Buy It Together
Current Item
QLA Clip Style Basket for Caleva, 10 Mesh, 316 SS; Serialized
QLA Clip Style Basket for Caleva, 10 Mesh, 316 SS; Serialized
4130128
₹48,500.00INR / Each
+
QLA Basket Holder, Universal Compatibility; 6 Position
4130170
₹39,100.00INR / Each
=
Total Price 87600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of 316 electropolished stainless steel All baskets are laser marked with unique serial numbers
  • 10 mesh
More About this Item
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Caleva test apparatus. All (except plastic suppository baskets) are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • Material316 stainless steel
  • Mesh Size10
  • DescriptionClip Style Basket for Caleva, 10 Mesh, 316 SS; Serialized