QLA Suppository Basket for Caleva; Plastic
Representative image only.
QLA Suppository Basket for Caleva; Plastic
| Mfr: BSKSUP-CA| Item: 4130193
QLA Baskets for dissolution testing with your Caleva apparatus
₹55,500.00INR / Each
₹55,500.00INR / Each
Buy It Together
Current Item
QLA Suppository Basket for Caleva; Plastic
QLA Suppository Basket for Caleva; Plastic
4130193
₹55,500.00INR / Each
+
QLA Basket Holder, Universal Compatibility; 6 Position
4130170
₹39,100.00INR / Each
=
Total Price 94600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of 316 electropolished stainless steel All baskets are laser marked with unique serial numbers
  • Suppository basket
More About this Item
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Caleva test apparatus. All (except plastic suppository baskets) are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • MaterialPlastic
  • DescriptionSuppository Basket for Caleva; Plastic