Pyrex Brand 36060 Funnels
Buchner, with fritted disc
Use these funnels in applications where filter paper would be attacked by the material being filtered. The disc should not be subjected to pressure differential in excess of one atmosphere. The top edge is beaded
79,700.00 - ₹110,000.00INR / Case of 6
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Disc Diameter (mm) Porosity
Availability Pricing
Pyrex 36060-30M Brand 36060 fritted funnel; 30 mL medium, case of 6 WW-34573-29 Mfr # 36060-30M
COMPARE
30 10 to 15 µm
₹79,700.00INR / Case of 6
Pyrex 36060-350F Brand 36060 fritted funnel; 350 mL fine, case of 3 WW-34573-37 Mfr # 36060-350F
COMPARE
80 4 to 5.5 µm
₹110,000.00INR / Case of 3