Pyrex Brand 3042 Cylinders
LIFETIME RED™, single metric scale
These single metric scale graduated cylinders are calibrated "to contain." The scale and figures marked in white enamel stand out clearly against the red color-band on these LIFETIME RED cylinders. The 10-mL size has a funnel top. Bumper guards are supplied with 100-mL through 2000-mL cylinders. The 100-mL size is made in accordance with ASTM E-133 and E-1272 and is ued in ASTM tests D-86 and D-216.
24,200.00 - ₹103,000.00INR / Case of 12
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Pyrex 3042-100 Brand 3042 cylinder; 100 mL, case of 12 WW-34547-22 Mfr # 3042-100
COMPARE
100 1 Glass
₹103,000.00INR / Case of 12
Pyrex 3042-1L Brand 3042 cylinder; 1000 mL, case of 1 WW-34547-24 Mfr # 3042-1L
COMPARE
1000 6 Glass
₹24,200.00INR / Case of 1
Pyrex 3042-2L Brand 3042 cylinder; 2000 mL, case of 1 WW-34547-25 Mfr # 3042-2L
COMPARE
2000 12 Glass
₹36,700.00INR / Case of 1