PVDF Magnetic Gamma Stable Stir Bars
Ideal for biopharmaceutical containers
  • Gamma Irradiated
  • Animal Derivative Component Free
  • Low Particulate Shedding
  • Meets USP Class VI
  • Autoclavable
Perfect for when your product needs to be both agitated and stay in suspension.
14,500.00 - ₹22,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Diameter (mm) Length (mm) Description
Availability Pricing
SaniSure SS-SB-0525-PVDF-G Magnetic Gamma Stable Stir Bars, 5 mm x 25 mm, Individually Bagged WW-04775-61 Mfr # SS-SB-0525-PVDF-G
COMPARE
5 25 Magnetic Gamma Stable Stir Bars, 5 mm x 25 mm, Individually Bagged
₹14,500.00INR / Each
SaniSure SS-SB-0625-PVDF-G Magnetic Gamma Stable Stir Bars, 6 mm x 25 mm, Individually Bagged WW-04775-63 Mfr # SS-SB-0625-PVDF-G
COMPARE
6 25 Magnetic Gamma Stable Stir Bars, 6 mm x 25 mm, Individually Bagged
₹15,300.00INR / Each
SaniSure SS-SB-0832-PVDF-G Magnetic Gamma Stable Stir Bars, 8 mm x 32 mm, Individually Bagged WW-04775-66 Mfr # SS-SB-0832-PVDF-G
COMPARE
8 32 Magnetic Gamma Stable Stir Bars, 8 mm x 32 mm, Individually Bagged
₹16,900.00INR / Each
SaniSure SS-SB-0851-PVDF-G Magnetic Gamma Stable Stir Bars, 8 mm x 51 mm, Individually Bagged WW-04775-67 Mfr # SS-SB-0851-PVDF-G
COMPARE
8 51 Magnetic Gamma Stable Stir Bars, 8 mm x 51 mm, Individually Bagged
₹19,460.00INR / Each
SaniSure SS-SB-1051-PVDF-G Magnetic Gamma Stable Stir Bars, 10 mm x 51 mm, Individually Bagged WW-04775-69 Mfr # SS-SB-1051-PVDF-G
COMPARE
10 51 Magnetic Gamma Stable Stir Bars, 10 mm x 51 mm, Individually Bagged
₹22,000.00INR / Each