PRO Scientific Cannabis Homogenizing Packages
Precision designed to provide reliable, reproducible results for cannabis testing and quality control
  • High-shear powerful and versatile handheld homogenizers
  • Compact size fits nicely in your hand
  • Variable-speed control from 5000 to 30,000 rpm
  • Built-in sensors prevent overheating—process samples for hours at a time
Each cannabis homogenizer package includes a durable, handheld homogenizer and a quick connect generator probe to provide quality cannabis sample preparation. The homogenizer is compact yet delivers 576 watts of high power with a variable speed range from 5000 to 30,000 rpm. A separate on/off switch allows for continuous processing.
Generator probes are constructed of autoclavable 316 stainless steel and PTFE. Basic packages include a saw-tooth bottom generator probe; deluxe packages include an open-slotted bottom generator probe. A three-step connection between the homogenizer and generator probe provides stability, durability, and longer generator probe life. Select a plus (+) model that includes a deflector head to help homogenize larger volumes and higher viscosity materials. Order an optional stand assembly to post mount the homogenizer unit.
792,000.00 - ₹1,014,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Sample Size (mL) Power (VAC) Tip Diameter (mm)
Availability Pricing
PRO Scientific Basic CBD Homogenizing Package; 115 VAC 1475200 Mfr # PK-01250B
COMPARE
3000 115 20
₹834,000.00INR / Each
PRO Scientific Basic CBD Homogenizing Package; 220 VAC 1475201 Mfr # PK-02250B
COMPARE
3000 220 20
₹792,000.00INR / Each
PRO Scientific Basic+ CBD Homogenizing Package; 115 VAC 1475202 Mfr # PK-01250BP
COMPARE
4000 115 20
₹898,000.00INR / Each
PRO Scientific Basic+ CBD Homogenizing Package; 220 VAC 1475203 Mfr # PK-02250BP
COMPARE
4000 220 20
₹854,000.00INR / Each
PRO Scientific Deluxe CBD Homogenizing Package; 115 VAC 1475204 Mfr # PK-01250D
COMPARE
5000 115 30
₹910,000.00INR / Each
PRO Scientific Deluxe CBD Homogenizing Package; 220 VAC 1475205 Mfr # PK-02250D
COMPARE
5000 220 30
₹958,000.00INR / Each
PRO Scientific Deluxe+ CBD Homogenizing Package; 115 VAC 1475206 Mfr # PK-01250DP
COMPARE
5000 115 30
₹964,000.00INR / Each
PRO Scientific Deluxe+ CBD Homogenizing Package; 220 VAC 1475207 Mfr # PK-02250DP
COMPARE
5000 220 30
₹1,014,000.00INR / Each
Showing 1- 8 of 8