Ohaus® Valor 3000 Portion Control Scales
Extreme performance in an extremely rugged stainless steel design
  • No exposed screws or edges—perfect for food testing and service environments
  • All stainless steel design with shock and overload protection for tough environments
  • NSF certified, USDA-AMS accepted, and HACCP-certified
These compact industrial balances offer an extremely rugged design for long-term performance in tough environments. Valor is NSF certified, USDA-AMS accepted, and supports HACCP-certified systems. No exposed screws or edges make these balances perfect for food testing and service environments and offer easy cleanup. Ohaus’ classic SmarText™-driven interface includes a brilliant backlit display and the Yes/No menu navigation offers fast, user-intuitive operation. User-adjustable stability settings and resolutions allow the Valor 3000 series to operate accurately in drafty and unstable environments.
162,000.00 - ₹331,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Ohaus V31XH4 Valor 3000 Portion Control Scale, 4 kg x 0.1 g 0103011 Mfr # V31XH4
COMPARE
4 6.615 100 to 220
₹331,000.00INR / Each
Ohaus V31XH202 Valor 3000Portion Control Scale, 200g x 0.01g 1101181 Mfr # V31XH202
COMPARE
0.2 0.44 115
₹243,000.00INR / Each
Ohaus V31XH2 Valor 3000Portion Control Scale, 2000g x 0.1g 1101182 Mfr # V31XH2
COMPARE
2 4 115
₹238,000.00INR / Each
Ohaus V31X3 Valor 3000Portion Control Scale, 3000Gx 1g 1101183 Mfr # V31X3
COMPARE
3 6.5 115
₹162,000.00INR / Each
Ohaus V31X6 Valor 3000Portion Control Scale, 6000g x 1g 1101184 Mfr # V31X6
COMPARE
6 13 115
₹162,000.00INR / Each
Ohaus V31XH402 Valor 3000Portion Control Scale, 400g x 0.01g 1161088 Mfr # V31XH402
COMPARE
0.4 0.88 100 to 240
₹331,000.00INR / Each