Ohaus Valor 2000 PW Portion Control Scales
A durable, rapid-response food processing scale that enhances safety, efficiency, and productivity even in harsh environments!
  • Thick polycarbonate keypad prevents damage from impacts and punctures
  • Flow Thru™ water-resistant design is rated IPX8, channeling liquids away from internal components
  • Fast response and large dual displays allow more than one operator to use the scale at the same time
  • NSF certified for food processing
  • Built-in rechargeable battery provides 50 hours continuous use
  • Spring-loaded power cord receptacle cover underneath scale protects connection from moisture
The Valor 2000 PW scales feature an ABS housing and removable stainless steel platform, and offer one mode for weighing. All models are designed with the Flow Thru™ water-resistant system, which channels liquids away from internal components and out through drain holes. This design includes silicone sealed internal components and connections to protect sensitive instrumentation from condensation.
170,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Ohaus V22PWE30 Valor 2000 PW Compact Bench Scale, 30 kg X 5 g 0103010 Mfr # V22PWE30T
COMPARE
0.5 1.1025 100 to 220
Call For Price
Ohaus V22PWE3T Valor 2000 PW Compact Bench Scale 3, 000g x 0.5G 1161126 Mfr # V22PWE3T
COMPARE
3 6 100 to 240
₹170,000.00INR / Each
Ohaus V22PWE15T Valor 2000 PW Compact Bench Scale 15, 000g x 2g 1161128 Mfr # V22PWE15T
COMPARE
15 30 100 to 240
₹170,000.00INR / Each