Ohaus® Triple-Beam Mechanical Balances
Classic mechanical balance with the durability and versatility to accommodate a range of lab applications
  • Spring-loaded zero adjust compensator
  • Magnetic damping speeds up weighing times
  • Select models feature a tare beam to accommodate containers up to 225 g
  • Select models come with Dial-O-Gram a convenient direct-reading dial calibration
  • Order weight set separately
34,300.00 - ₹92,200.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g) Description
Availability Pricing
Ohaus 710-TO Triple-Beam Balance with Tare and Deep Stainless Steel Pan, 610g x 0.1g WW-01006-01 Mfr # 710-T0
COMPARE
610 0.1 Triple-Beam Balance with Tare and Deep Stainless Steel Pan, 610g x 0.1g
₹75,800.00INR / Each
Ohaus 760-00 Triple-Beam Balance with Tare and Stainless Steel Pan, 610g x 0.1g, WW-01012-00 Mfr # 760-00
COMPARE
610 0.1 Triple-Beam Balance, 610g x 0.1g, Stainless Steel Weighing Plate
₹64,100.00INR / Each
Ohaus 1610-00 Dial-O-Gram Triple Beam Balance with Tare and SS Pan, 610g x 0.1g SKU 1110019 WW-11100-19 Mfr # 1610-00
COMPARE
610 0.1 Dial-O-Gram Triple Beam Balance with Tare and SS Pan, 610g x 0.1g
₹92,200.00INR / Each
Ohaus 1650-00 Dial-O-Gram Triple-Beam Balance, Stainless Steel Pan, 610g x 0.1g WW-11100-22 Mfr # 1650-00
COMPARE
610 0.1 Dial-O-Gram Triple-Beam Balance, Stainless Steel Pan, 610g x 0.1g
₹80,000.00INR / Each
Ohaus 750-S0 Triple-Beam Balance with Stainless Steel Pan, 610g x 0.1g WW-11100-26 Mfr # 750-S0
COMPARE
610 0.1 Triple-Beam Balance with Tare and Stainless Steel Pan, 610g x 0.1g
₹34,300.00INR / Each
Ohaus 720-00 Triple-Beam Balance with Scoop, 610g x 0.1g WW-11610-71 Mfr # 720-00
COMPARE
610 0.1 Triple-Beam Balance with Scoop, 610g x 0.1g
₹72,800.00INR / Each
Ohaus 720-S0 Triple-Beam Balance with Metal Scoop, 610g x 0.1g WW-11610-72 Mfr # 720-S0
COMPARE
610 0.1 Triple-Beam Balance with Metal Scoop, 610g x 0.1g
₹84,500.00INR / Each
Showing 1- 7 of 7