Ohaus Ranger 7000 Compact Bench Scales
Complex industrial applications made simple
  • Ten advanced features minimize the need for manual calculations
  • IP54-rated cast-metal housing and sealed metal indicator withstand the harshest conditions
  • Store 300 sets of weighing, check, counting, or filling data, and 30 sets of formulation data
  • SmarText 2.0 interface is user-friendly, reducing training time for operators
  • Standard RS-232 and USB ports export GLP/GMP data to PC or printer, with other communication modules available separately
  • Lights and audible signals make check weighing quick and easy
  • Level and stability indicators ensure accurate results
  • Overload/underload indicators and menu-lock switch provide security
  • Modular design allows indicator to be mounted to a wall, bench or optional tower
  • With optional remote base and external scale platform, results can be viewed from both platforms simultaneously
  • Optional discrete I/O interface provides control via peripheral devices
  • Choose between standard and high resolution models to suit your application''s requirements for precision
The Ohaus® Ranger® 7000 compact bench scale simplifies even the most complex industrial applications utilizing the SmarText 2.0 interface, internal memory, and a versatile modular design. The Legal for Trade certifications and full GLP/GMP data output make this scale an ideal choice for regulated industries.
587,000.00 - ₹1,087,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Ohaus R71MD3 Ranger 7000 Compact Bench Scale 3kg x 0.00005kg 9045278 Mfr # R71MD3
COMPARE
3 6 100 to 240
₹587,000.00INR / Each
Ohaus R71MD6 Ranger 7000 Compact Bench Scale 6kg x 0.0001kg 9045279 Mfr # R71MD6
COMPARE
6 15 100 to 240
₹587,000.00INR / Each
Ohaus R71MD15 Ranger 7000 Compact Bench Scale 15kg x 0.0002kg 9045280 Mfr # R71MD15
COMPARE
15 30 100 to 240
₹587,000.00INR / Each
Ohaus R71MD35 Ranger 7000 Compact Bench Scale 35kg x 0.0005kg 9045281 Mfr # R71MD35
COMPARE
35 70 100 to 240
₹587,000.00INR / Each
Ohaus R71MHD3 Ranger 7000 High Res Compact Bench Scale 3kg x 0.00001kg 9045282 Mfr # R71MHD3
COMPARE
3 6 100 to 240
₹1,087,000.00INR / Each
Ohaus R71MHD6 Ranger 7000 High Res Compact Bench Scale 6kg x 0.00002kg 9045283 Mfr # R71MHD6
COMPARE
6 15 100 to 240
₹1,034,000.00INR / Each
Ohaus R71MHD15 Ranger 7000 High Res Compact Bench Scale 15kg x 0.0001kg 9045284 Mfr # R71MHD15
COMPARE
15 30 100 to 240
Call For Price
Ohaus R71MHD35 Ranger 7000 High Res Compact Bench Scale 35kg x 0.0001kg 9045285 Mfr # R71MHD35
COMPARE
35 70 100 to 240
Call For Price
Showing 1- 8 of 8